missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ R 268712
GREENER_CHOICE

Product Code. 18729228
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18729228

Brand: Tocris Bioscience™ 5288/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective inhibitor of TGF-βRI

R 268712 is a potent and selective inhibitor of the TGF-β type I receptor/ALK5 (IC50 = 2.5 nM). Exhibits ∽5000-fold selectivity over p38 MAPK. Suppresses development of renal fibrosis in a unilateral ureteral obstruction model. Also reduces proteinuria, glomerulosclerosis and improves renal function in an anti-Thy1 glomerulonephritis model. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type R 268712
Purity >98%
Molecular Weight (g/mol) 363.39
Inhibitors R 268712

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.