missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Amino-1-benzylpiperidine, 97%

Product Code. 15459157
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459157 1 g 1g
15469157 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459157 Supplier Thermo Scientific Alfa Aesar Supplier No. H52425.03
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 168466-84-0 | C12H18N2 | 190.29 g/mol

(R)-3-Amino-1-benzylpiperidine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-3-Amino-1-benzylpiperidine is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 168466-84-0
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD03093374
InChI Key HARWNWOLWMTQCC-GFCCVEGCSA-N
Synonym r-3-amino-1-benzylpiperidine, r-1-benzylpiperidin-3-amine, r-3-amino-1-benzyl-piperidine, 3r-1-benzylpiperidin-3-amine, r-1-benzyl-3-aminopiperidine, 3-piperidinamine, 1-phenylmethyl-, 3r, s-3-amino-n-benzylpiperidine, pubchem11493, r-n-benzyl-3-aminopiperidine, r-1-benzyl-piperidin-3-ylamine
PubChem CID 854130
IUPAC Name (3R)-1-benzylpiperidin-3-amine
SMILES C1CC(CN(C1)CC2=CC=CC=C2)N

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 190.29
Percent Purity 97%
Chemical Name or Material (R)-3-Amino-1-benzylpiperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.