missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-3-Amino-1-Boc-piperidine, 98%

Product Code. 15453567
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453567 5 g 5g
15443567 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453567 Supplier Thermo Scientific Alfa Aesar Supplier No. H26937.06
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 188111-79-7 | C10H20N2O2 | 200.282 g/mol

(R)-(-)-3-Amino-1-Boc-piperidine is used as an intermediate in the manufacture of various substances such as pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-3-Amino-1-Boc-piperidine is used as an intermediate in the manufacture of various substances such as pharmaceuticals.

Solubility
Immiscible with water.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188111-79-7
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD03094717
InChI Key AKQXKEBCONUWCL-MRVPVSSYSA-N
Synonym r-1-boc-3-aminopiperidine, r-tert-butyl 3-aminopiperidine-1-carboxylate, r-3-amino-1-n-boc-piperidine, tert-butyl 3r-3-aminopiperidine-1-carboxylate, r-1-boc-3-amino-piperidine, r-3-amino-1-boc-piperidine, s-n-boc-3-aminopiperidine, r-3-amino-piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 3r, r-1-tert-butoxycarbonyl-3-aminopiperidine
PubChem CID 1501974
IUPAC Name tert-butyl (3R)-3-aminopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)N

Specifications

Refractive Index 1.473
Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Immiscible with water.
Optical Rotation −28° (Neat)
Formula Weight 200.28
Percent Purity 98%
Chemical Name or Material (R)-(-)-3-Amino-1-Boc-piperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.