missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Amino-3-phenylpropan-1-ol, 95%, 98% ee, Thermo Scientific™

Product Code. 10272712 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
500mg
Product Code. Quantity Packaging unitSize
10272712 500 mg Glass bottle 500mg
10180802 1 g Glass bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10272712

Brand: Thermo Scientific Acros 363125000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 170564-98-4
MDL Number MFCD01311791
InChI Key SEQXIQNPMQTBGN-SECBINFHSA-N
Synonym r-1-phenyl-3-propanolamine, r-3-amino-3-phenylpropan-1-ol, 3r-3-amino-3-phenylpropan-1-ol, r-beta-phenylalaninol, r-3-amino-3-phenyl-1-propanol, r-3-phenyl-beta-alaninol, r-3-amino-3-phenyl-propan-1-ol, r--phenylalaninol, pubchem13853
PubChem CID 7016858
IUPAC Name (3R)-3-amino-3-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)C(CCO)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White to Yellow or Yellow
Enantiomeric Excess 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C9H13NO
Quantity 500 mg
Molecular Weight (g/mol) 151.209
Formula Weight 151.21
Percent Purity ≥94%
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material (R)-3-Amino-3-phenylpropan-1-ol, 95%, (98% e.e.)
Show More Show Less
compliance-icons
Product Identifier
  • (R)-3-Amino-3-phenylpropan-1-ol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.