missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-3-Butyn-2-ol, 99%

Product Code. 11329535
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11329535 250 mg 250mg
11309535 1 g 1g
11319535 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11329535 Supplier Thermo Scientific Alfa Aesar Supplier No. L18880.MD
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 42969-65-3 | C4H6O | 70.091 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42969-65-3
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.091
MDL Number MFCD00211237
InChI Key GKPOMITUDGXOSB-SCSAIBSYSA-N
Synonym r-+-3-butyn-2-ol, r-3-butyn-2-ol, 2r-but-3-yn-2-ol, r-but-3-yn-2-ol, 2r-3-butyn-2-ol, 3-butyn-2-ol, 2r, r-+-1-butyn-3-ol, +-3-butyn-2-ol, pubchem20211, r---3-butyne-2-ol
PubChem CID 638102
IUPAC Name (2R)-but-3-yn-2-ol
SMILES CC(C#C)O

Specifications

Density 0.891
Boiling Point 108°C to 109°C
Flash Point 28°C (82°F)
Refractive Index 1.426
Quantity 250 mg
UN Number UN2929
Beilstein 3587272
Optical Rotation +47° (Neat)
Formula Weight 70.09
Percent Purity 99%
Chemical Name or Material (R)-(+)-3-Butyn-2-ol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.