missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (R)-(+)-3-Chloro-1-phenyl-1-propanol, 98%

Product Code. 15461868
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15461868 250 mg 250mg
15441868 1 g 1g
15451868 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15461868 Supplier Alfa Aesar™ Supplier No. H55567.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 100306-33-0
Molecular Formula C9H11ClO
Molecular Weight (g/mol) 170.636
MDL Number MFCD00075128
InChI Key JZFUHAGLMZWKTF-SECBINFHSA-N
Synonym r-3-chloro-1-phenylpropan-1-ol, r-+-3-chloro-1-phenyl-1-propanol, r-+-3-chloro-1-phenylpropanol, 1r-3-chloro-1-phenylpropan-1-ol, 1r-3-chloro-1-phenyl-propan-1-ol, r-3-chloro-1-phenyl-1-propanol, r-3-chloro-1-phenyl-propan-1-ol, r-3-chloro-1-phenylpropanol, pubchem5738
PubChem CID 642409
IUPAC Name (1R)-3-chloro-1-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)C(CCCl)O

Specifications

Melting Point 58°C to 60°C
Quantity 250 mg
Beilstein 5250766
Optical Rotation +26° (c=1 in Chloroform)
Formula Weight 170.64
Percent Purity 98%
Chemical Name or Material (R)-(+)-3-Chloro-1-phenyl-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.