missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Hydroxyisobutyric Acid Methyl Ester, TRC

Product Code. 30408438
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30408438 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30408438 Supplier Toronto Research Chemicals Supplier No. TRCH94355825G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-3-Hydroxyisobutyric Acid Methyl Ester
CAS 72657-23-9
Molecular Formula C5 H10 O3
Quantity 25 g
Synonym Propanoic acid, 3-hydroxy-2-methyl-, methyl ester, (2R)-, Propanoic acid, 3-hydroxy-2-methyl-, methyl ester, (R)-, (-)-(R)-Roche ester, (-)-Methyl β-hydroxyisobutyrate, (2R)-3-Hydroxy-2-methylpropionic acid methyl ester, (R)-(-)-3-Hydroxy-2-methylpropionic acid methyl ester, (R)-3-Hydroxy-2-methylpropionic acid methyl ester, (R)-3-Hydroxy-2-methylpropionic acid methyl ester, (R)-Methyl 3-hydroxy-2-methylpropanoate, (R)-Roche ester, (R)-Roche ester, Methyl (2R)-3-hydroxy-2-methylpropanoate, Methyl (2R)-3-hydroxy-2-methylpropionate, Methyl (R)-(-)-3-hydroxy-2-methylpropionate, Methyl (R)-(-)-3-hydroxy-2-methylpropionate, Methyl (R)-3-hydroxy-2-methylpropanoate, Methyl (R)-3-hydroxy-2-methylpropanoate, Methyl (R)-3-hydroxy-2-methylpropionate, Methyl (R)-3-hydroxyisobutanoate, Methyl (R)-β-hydroxyisobutyrate, R-(-)-Roche ester
InChI Formula InChI=1S/C5H10O3/c1-4(3-6)5(7)8-2/h4,6H,3H2,1-2H3/t4-/m1/s1
SMILES COC(=O)[C@H](C)CO
IUPAC Name methyl (2R)-3-hydroxy-2-methylpropanoate
Molecular Weight (g/mol) 118.13
Formula Weight 118.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.