missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-3-Hydroxytetrahydrofuran, 98%, Thermo Scientific™

Product Code. 15452768
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15452768 250 mg 250mg
15432768 1 g 1g
15442768 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15452768 Supplier Thermo Scientific Alfa Aesar Supplier No. H56307.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86087-24-3
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00067101
InChI Key XDPCNPCKDGQBAN-SCSAIBSYSA-N
Synonym r---3-hydroxytetrahydrofuran, r-tetrahydrofuran-3-ol, r-3-hydroxytetrahydrofuran, 3r-oxolan-3-ol, 3r-tetrahydrofuran-3-ol, r-+-3-hydroxytetrahydrofuran, r-tetrahydro-furan-3-ol, 3-furanol, tetrahydro-, 3r, pubchem7071, 2-furanyl,tetrahydro-3-hydroxy-, r-9ci
PubChem CID 641512
IUPAC Name (3R)-oxolan-3-ol
SMILES C1COCC1O

Specifications

Density 1.097
Quantity 250 mg
Refractive Index 1.45
Boiling Point 181°C
Formula Weight 88.11
Percent Purity 98%
Chemical Name or Material (R)-(-)-3-Hydroxytetrahydrofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.