missing translation for 'onlineSavingsMsg'
Learn More

(R)-4-Boc-morpholine-3-carboxylic acid, 97%

Product Code. 15421949
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15421949 250 mg 250mg
15411949 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15421949 Supplier Thermo Scientific Alfa Aesar Supplier No. H64654.MD
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 869681-70-9 | C10H17NO5 | 231.248 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 869681-70-9
Molecular Formula C10H17NO5
Molecular Weight (g/mol) 231.248
MDL Number MFCD04114909
InChI Key KVXXEKIGMOEPSA-SSDOTTSWSA-N
Synonym r-4-tert-butoxycarbonyl morpholine-3-carboxylic acid, r-4-boc-morpholine-3-carboxylic acid, 4-boc-3 r-morpholinecarboxylic acid, r-morpholine-3,4-dicarboxylic acid 4-tert-butyl ester, r-n-boc-morpholine-3-carboxylic acid, r-4-n-boc-3-morpholinecarboxylic acid, r-4-boc-morpholine-3-carboxylicacid, 3r-4-tert-butoxycarbonyl morpholine-3-carboxylic acid, 3,4-morpholinedicarboxylic acid, 4-1,1-dimethylethyl ester, 3r, ambotzbaa5270
PubChem CID 1512538
IUPAC Name (3R)-4-[(2-methylpropan-2-yl)oxycarbonyl]morpholine-3-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCOCC1C(=O)O

Specifications

Melting Point 181°C
Color White
UN Number UN2811
Beilstein 10254221
Quantity 250 mg
Formula Weight 231.25
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (R)-4-Boc-morpholine-3-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.