missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-4-Methyl-2-pentanol, 99%

Product Code. 11349535
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11349535

Brand: Thermo Scientific Alfa Aesar L18881.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16404-54-9 | C6H14O | 102.177 g/mol

For synthesis of optically active products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16404-54-9
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD03093077
InChI Key WVYWICLMDOOCFB-ZCFIWIBFSA-N
Synonym r---4-methyl-2-pentanol, 2r-4-methylpentan-2-ol, r-4-methyl-2-pentanol, r-4-methylpentan-2-ol, r---4-methylpentan-2-ol, 4-methyl r-2-pentanol, 2r-4-methyl-2-pentanol, r---methylpentan-2-ol, r--4-methyl-2-pentanol, 2-pentanol, 4-methyl-, 2r
PubChem CID 6999987
IUPAC Name (2R)-4-methylpentan-2-ol
SMILES CC(C)CC(C)O

Specifications

Density 0.802
Boiling Point 130°C to 131°C
Flash Point 41°C (105°F)
Odor Slightly Sweetish
Refractive Index 1.411
Quantity 250 mg
UN Number UN2053
Beilstein 1718992
Optical Rotation −21° (Neat)
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material (R)-(-)-4-Methyl-2-pentanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.