missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-4-Methylmandelic acid, ChiPros∣r, 98%, ee 97+%

Product Code. 11415730
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415730 5 g 5g
11405730 1 g 1g
11425730 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11415730 Supplier Thermo Scientific Alfa Aesar Supplier No. H25953.06
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31284-89-6 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31284-89-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00798433,MFCD00961860
InChI Key SFGURAWGCAPHON-UHFFFAOYNA-N
Synonym 2-hydroxy-2-4-methylphenyl acetic acid, 4-methylmandelic acid, p-methyl mandelic acid, 2-hydroxy-2-p-tolyl acetic acid, hydroxy 4-methylphenyl acetic acid, hydroxy-p-tolyl-acetic acid, p-methylmandelic acid, 4-methylmaldelic acid, p-methyl-mandelic acid, acmc-1afsj
PubChem CID 12458823
IUPAC Name (2R)-2-hydroxy-2-(4-methylphenyl)acetic acid
SMILES CC1=CC=C(C=C1)C(O)C(O)=O

Specifications

Melting Point 130°C to 133°C
Quantity 5 g
Sensitivity Light sensitive
Optical Rotation −154° (c=1 in Methanol)
Formula Weight 166.18
Percent Purity 98%
Assay ee ≥97%
Chemical Name or Material (R)-(-)-4-Methylmandelic acid, ChiPros

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.