missing translation for 'onlineSavingsMsg'
Learn More

Atago™ R–5000 Hand-Held Refractometer

Product Code. 10420705
Change view
Click to view available options
Refractive Index:
1.333 to 1.520
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Refractive Index Unit Size
10420705 1.333 to 1.520 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10420705 Supplier Atago™ Supplier No. 2350
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for simple, quick and easy measurement of refractive index. Atago™ R–5000 Hand-Held Refractometer features a 3-stage switching scale.

  • Refractive index scale
  • Three-stage, switching scale range

Specifications

Type Handheld Refractometer
Refractive Index 1.333 to 1.520
Model R–5000
For Use With (Application) Simple, quick, and easy measurement of refractive index
Weight (Metric) 600 g
Dimensions (L x W x H) 4 x 4 x 20 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.