missing translation for 'onlineSavingsMsg'
Learn More

R(-)-Alpha-Methyl Histamine Dihydrochloride, TRC

Product Code. 30135935
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135935 5 mg 5mg
30134875 25 mg 25mg
30007526 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135935 Supplier Toronto Research Chemicals Supplier No. TRCM3119905MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75614-89-0
Molecular Formula C6 H11 N3 . 2 Cl H
Molecular Weight (g/mol) 198.09
Synonym (αR)-α-Methyl-1H-imidazole-4-ethanamine Dihydrochloride, R(-)-α-Methyl-1H-imidazole-5-ethanamine Dihydrochloride;
IUPAC Name (2R)-1-(1H-imidazol-4-yl)propan-2-amine;dihydrochloride
SMILES Cl.Cl.C[C@@H](N)Cc1c[nH]cn1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C6H11N3.2ClH/c1-5(7)2-6-3-8-4-9-6;;/h3-5H,2,7H2,1H3,(H,8,9);2*1H/t5-;;/m1../s1
Formula Weight 197.0487
Chemical Name or Material R(-)-alpha-Methyl Histamine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.