missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Alpha-Methylbenzylamine (>85%), TRC

Product Code. 30179287
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179287 10 g 10g
30179007 50 g 50g
30179145 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179287 Supplier Toronto Research Chemicals Supplier No. TRCM38880010G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3886-69-9
Molecular Formula C8 H11 N
Molecular Weight (g/mol) 121.18
Synonym Benzenemethanamine, α-methyl-, (αR)-, Benzenemethanamine, α-methyl-, (R)-, Benzylamine, α-methyl-, (R)-(+)- (8CI), Benzylamine, α-methyl-, d- (4CI), (αR)-α-Methylbenzenemethanamine, (+)-(R)-1-Phenethylamine, (+)-(R)-1-Phenylethylamine, (+)-(R)-α-Methylbenzylamine, (+)-(R)-α-Phenylethylamine, (+)-1-Amino-1-phenylethane, (+)-1-Phenethylamine, (+)-1-Phenylethan-1-amine, (+)-1-Phenylethylamine, (+)-Phenylethylamine, (+)-[(1R)-1-Phenylethyl]amine, (+)-α-Methylbenzenemethanamine, (+)-α-Methylbenzylamine, (+)-α-Phenethylamine, (+)-α-Phenylethylamine, (1R)-1-Phenyl-1-ethanamine, (1R)-1-Phenylethan-1-amine, (1R)-1-Phenylethanamine, (1R)-1-Phenylethanamine, (1R)-1-Phenylethylamine, (1R)-Phenylethylamine, (R)-(+)-1-Methylbenzylamine, (R)-(+)-1-Phenethylamine, (R)-(+)-1-Phenylethanamine, (R)-(+)-1-Phenylethylamine, (R)-(+)-Alpha-methylbenzylamine, (R)-(+)-Phenethylamine, (R)-(+)-α-Methylbenzylamine, (R)-(+)-α-Phenethylamine, (R)-(+)-α-Phenylethylamine, (R)-1-Amino-1-phenylethane, (R)-1-Aminoethylbenzene, (R)-1-Phenethylamine, (R)-1-Phenyl-1-ethanamine, (R)-1-Phenylethanamine, (R)-1-Phenylethylamine, (R)-Alpha-methylbenzylamine, (R)-Methylbenzylamine, (R)-Phenethylamine, (R)-Phenylethylamine, (R)-Phenylethylamine, (R)-α-Aminoethylbenzene, (R)-α-Methylbenzenemethanamine, (R)-α-Methylbenzylamine, (R)-α-Phenethylamine, (R)-α-Phenylethylamine, D-(+)-1-Phenylethylamine, D-(+)-α-Methylbenzylamine, D-(+)-α-Phenylethylamine, D-1-Phenylethylamine, D-Phenethylamine, D-α-Methylbenzylamine, D-α-Phenethylamine, R-(+)-Methylbenzylamine, R-(+)-Phenylethylamine, d-1-Phenylethylamine, Levetiracetam Imp. E (EP)
IUPAC Name (1R)-1-phenylethanamine
SMILES C[C@@H](N)c1ccccc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H11N/c1-7(9)8-5-3-2-4-6-8/h2-7H,9H2,1H3/t7-/m1/s1
Formula Weight 121.0891
Chemical Name or Material (R)-(+)-a-Methylbenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.