missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-Carvone-4,4,6,6-d4, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | (R)-(-)-Carvone 4,4,6,6-d4 |
| Molecular Formula | C10 D4 H10 O |
| Quantity | 0.5 g |
| Synonym | (R)-(-)-Carvone-4,4,6,6-d4 |
| InChI Formula | InChI=1 S/C10H14O/c1-7(2)9-5-4-8(3)10(11)6-9/h4,9 H,1,5-6H2,2-3H3/t9-/m1/s1/i5D2,6D2 |
| SMILES | [2 H]C1([2 H])C=C(C)C(=O)C([2 H])([2 H])[C@@H]1 C(=C)C |
| IUPAC Name | (5 R)-4,4,6,6-tetradeuterio-2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-one |
| Molecular Weight (g/mol) | 154.242 |
| Formula Weight | 154.13 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction