missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Carvone, 98%

Product Code. 11441608
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441608 50 g 50g
11451608 250 g 250g
11431608 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11441608 Supplier Thermo Scientific Alfa Aesar Supplier No. A13900.18
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6485-40-1 | C10H14O | 150.22 g/mol

(R)-(-)-Carvone is used in the flavor and food industry such as chewing gum additives. It is used in air freshening products such as in essential oils as well as in aromatherapy and alternative medicine. It is used to prepare carvomenthol, carvomenthone, dihydrocarvone, carveol and limonene. It reacts with lithium dimethylcuprate to place a methyl group trans to the isopropenyl group with good stereoselectivity. It is also used as a mosquito repellent and prevent premature sprouting of potatoes. Further, it is employed as an important starting material for the synthesis of enantiopure (R)-(+)-3-methyl-6-isopropenyl-cyclohept-3-enone-1 and (4S,6R,7R)-trihydroxy-1-octyne derivatives. It is utilized as a vital raw material for the asymmetric total synthesis of natural products. In addition to this, it is used as a chiral starting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-Carvone is used in the flavor and food industry such as chewing gum additives. It is used in air freshening products such as in essential oils as well as in aromatherapy and alternative medicine. It is used to prepare carvomenthol, carvomenthone, dihydrocarvone, carveol and limonene. It reacts with lithium dimethylcuprate to place a methyl group trans to the isopropenyl group with good stereoselectivity. It is also used as a mosquito repellent and prevent premature sprouting of potatoes. Further, it is employed as an important starting material for the synthesis of enantiopure (R)-(+)-3-methyl-6-isopropenyl-cyclohept-3-enone-1 and (4S,6R,7R)-trihydroxy-1-octyne derivatives. It is utilized as a vital raw material for the asymmetric total synthesis of natural products. In addition to this, it is used as a chiral starting material.

Solubility
Miscible with alcohol, ether, chloroform, propylene glycol and mineral oils. Immiscible with water and glycerol.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6485-40-1
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00001578
InChI Key ULDHMXUKGWMISQ-SECBINFHSA-N
Synonym --carvone, l-carvone, r---carvone, l--carvone, r-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, 4r-carvone, levo-carvone, --p-mentha-6,8-dien-2-one, --r-carvone
PubChem CID 439570
ChEBI CHEBI:15400
SMILES CC(=C)[C@@H]1CC=C(C)C(=O)C1

Specifications

Density 0.959
Boiling Point 228°C to 230°C
Flash Point 93°C (199°F)
Assay Percent Range 98%
Refractive Index 1.499
Quantity 50 g
Beilstein 2206714
Solubility Information Miscible with alcohol,ether,chloroform,propylene glycol and mineral oils. Immiscible with water and glycerol.
IUPAC Name (5R)-2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-one
Formula Weight 150.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (R)-(-)-Carvone
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(-)-Carvone
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: (-)-Carvone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.