missing translation for 'onlineSavingsMsg'
Learn More

(R)-cis-5Xi-Methyl Atracurium Dibesylate, TRC

Product Code. 30010475
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010475 5 mg 5mg
30009865 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010475 Supplier Toronto Research Chemicals Supplier No. TRCM2883205MG
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193104-82-3
Molecular Formula C54H74N2O12•2(C6H5O3S)
Molecular Weight (g/mol) 943.17215717
Synonym (1R,1'R,2R,2'R)-2,2'-[(3-Methyl-1,5-pentanediyl)bis[oxy(3-oxo-3,1-propanediyl)]]bis[1-[(3,4-dimethoxyphenyl)methyl]-1,2,3,4-tetrahydro-6,7-dimethoxy-2-methyl-Isoquinolinium Dibenzenesulfonate, Atracurium EP Impurity K
SMILES O=C(OCCCCC(C)OC(CC[N@+]1(C)[C@H](CC2=CC(OC)=C(OC)C=C2)C(C=C(OC)C(OC)=C3)=C3CC1)=O)CC[N@+]4(C)[C@H](CC5=CC=C(OC)C(OC)=C5)C6=CC(OC)=C(OC)C=C6CC4

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C54H74N2O12/c1-36(68-54(58)22-26-56(3)24-20-40-33-50(64-9)52(66-11)35-42(40)44(56)29-38-16-18-46(60-5)48(31-38)62-7)14-12-13-27-67-53(57)21-25-55(2)23-19-39-32-49(63-8)51(65-10)34-41(39)43(55)28-37-15-17-45(59-4)47(30-37)61-6/h15-18,30-36,43-44H,12-14,19-29H2,1-11H3/q+2/t36?,43-,44-,55-,56-/m1/s1
Chemical Name or Material (R)-cis-5xi-Methyl Atracurium Dibesylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.