missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (R)-Citalopram oxalate
GREENER_CHOICE

Product Code. 18707978
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18707978 10 mg 10mg
18743989 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18707978 Supplier Tocris Bioscience™ Supplier No. 5763/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Enantiomer of escitalopram oxalate (Cat. No. 1427)

(R)-Citalopram oxalate is an enantiomer of escitalopram oxalate . Postulated to allosterically modulate the affinity of the S-enantiomer.

Racemate and S-enantiomer also available.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (-)-(R)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile oxalate
CAS 219861-53-7
Quantity 10 mg
Target 5-HT Transporter Inhibitors
Molecular Formula C20H21FN2O.C2H2O4
Formula Weight 414.43
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.