missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ (R)-Citalopram oxalate

Description
(R)-Citalopram oxalate is an enantiomer of escitalopram oxalate . Postulated to allosterically modulate the affinity of the S-enantiomer.
Racemate and S-enantiomer also available.
Specifications
Specifications
| Chemical Name or Material | (-)-(R)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile oxalate |
| CAS | 219861-53-7 |
| Quantity | 10 mg |
| Target | 5-HT Transporter Inhibitors |
| Molecular Formula | C20H21FN2O.C2H2O4 |
| Formula Weight | 414.43 |
| Purity | >98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?