missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ (R)-CPP

Product Code. 30001068
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001068 10 mg 10mg
18074432 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001068 Supplier R&D Systems™ Supplier No. 0247/10
  • ONLINE EXCLUSIVE PRICE
    573.97 €
    514.50 € / 10mg
    Save 59.47 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent NMDA antagonist; more active enantiomer of (RS)-CPP

Highly potent NMDA antagonist; more active isomer. Shows some selectivity for GluN2A (formally NR2A) containing receptors (Ki values are 0.041, 0.27, 0.63 and 1.99 μM for inhibition of GluN2A-, GluN2B-, GluN2C- and GluN2D-containing recombinant NMDA receptors respectively).

Racemate also available.

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126453-07-4
Molecular Formula C8H17N2O5P

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 252.21
Purity >98%
Molecular Weight (g/mol) 252.207
Chemical Name or Material (R)-CPP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.