missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ABT 089 Dihydrochloride

Description
ABT 089 dihydrochloride is a high affinity and selective α4β2 partial agonist (Ki = 16 nM). Exhibits >1000-fold and >10,000-fold selectivity for α4β2 over α1β1γ1 and α7 receptors respectively. Neuroprotective against glutamate-induced toxicity in rat cortical neurons in vitro. Enhances cognitive performance in vivo. Orally bioavailable and brain penetrant.
Specifications
Specifications
| Chemical Name or Material | ABT 089 dihydrochloride |
| CAS | 161416-61-1 |
| Synonym | 2-Methyl-3-[(2S)-pyrrolidinylmethoxy]pyridine dihydrochloride |
| Quantity | 50 mg |
| Solubility | Soluble to 100mM in water and to 20mM in DMSO |
| Molecular Formula | C11H16N2O·2HCl |
| Formula Weight | 265.18 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?