Learn More
Tocris Bioscience™ Aripiprazole
High affinity D2 and 5-HT1A receptor partial agonist; also 5-HT2A antagonist
Brand:  Tocris Bioscience™ 5584/50
Encompass
Product Code. 15766946
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Aripiprazole is a high affinity dopamine D2 and 5-HT1A receptor partial agonist (Ki values are ∽1.64 and 5.59 nM respectively). Also 5-HT2A antagonist (Ki = 8.7 nM). Suppresses morphine-induced emesis in vivo. Atypical antipsychotic.
Specifications
Specifications
Aripiprazole
7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydro-2(1H)-quinolinone
Soluble to 100mM in DMSO
Bioactive Small Molecule
129722-12-9
50 mg
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Product Title
Select an issue
Research Use Only. Not For Human or Veterinary Use