missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Aripiprazole

Product Code. 18799937
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18799937 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18799937 Supplier R&D Systems™ Supplier No. 5584/50
  • ONLINE EXCLUSIVE PRICE
    179.04 €
    160.65 € / 50mg
    Save 18.39 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. High affinity D2 and 5-HT1A receptor partial agonist; also 5-HT2A antagonist

Aripiprazole is a high affinity dopamine D2 and 5-HT1A receptor partial agonist (Ki values are ∽1.64 and 5.59 nM respectively). Also 5-HT2A antagonist (Ki = 8.7 nM). Suppresses morphine-induced emesis in vivo. Atypical antipsychotic.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aripiprazole
CAS 129722-12-9
Synonym 7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydro-2(1H)-quinolinone
Quantity 50 mg
Solubility Soluble to 100mM in DMSO
Product Line Tocris
Molecular Formula C23H27Cl2N3O2
Formula Weight 448.39
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >99%
Show More Show Less

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.