missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ASP 7663

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ASP 7663 is a selective TRPA1 activator (EC50 values are ∽0.5 μM in human, mouse and rat). Displays little or no affinity at more than 60 other related receptors, channels and enzymes. Stimulates 5-HT release from QGP-1 cells. Exhibits an abdominal analgesic effect in vivo. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | ASP 7663 |
| CAS | 1190217-35-6 |
| Synonym | (2E)-2-[7-Fluoro-1,2-dihydro-1-(2-methylpropyl)-2-oxo-3H-indol-3-ylidene]acetic acid |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO and to 100mM in ethanol |
| Molecular Formula | C14H14FNO3 |
| Formula Weight | 263.26 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction