missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ASP 7663
GREENER_CHOICE

Product Code. 18730398
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18730398

Brand: Tocris Bioscience™ 5178/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective TRPA1 activator

ASP 7663 is a selective TRPA1 activator (EC50 values are ∽0.5 μM in human, mouse and rat). Displays little or no affinity at more than 60 other related receptors, channels and enzymes. Stimulates 5-HT release from QGP-1 cells. Exhibits an abdominal analgesic effect in vivo. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material ASP 7663
CAS 1190217-35-6
Synonym (2E)-2-[7-Fluoro-1,2-dihydro-1-(2-methylpropyl)-2-oxo-3H-indol-3-ylidene]acetic acid
Quantity 50 mg
Solubility Soluble to 100mM in DMSO and to 100mM in ethanol
Molecular Formula C14H14FNO3
Formula Weight 263.26
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.