missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ C 87

Product Code. 18797658
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18797658

Brand: Tocris Bioscience™ 5484/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

TNF-α inhibitor

C 87 is a TNF-α inhibitor (Kd = 110 nM for hTNF-α). Inhibits TNF-α-induced cytotoxicity in L929 fibroblast cells. Attenuates inflammation and increases survival in a mouse hepatitis model.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material C 87
CAS 332420-90-3
Synonym 3-Phenyl-1-(4-phenyl-2-thiazolyl)-1H-pyrazole-4,5-dione 4-[2-(4-chloro-3-nitrophenyl)hydrazone]
Quantity 10 mg
Solubility Soluble to 20mM in DMSO
Molecular Formula C24H15ClN6O3S
Formula Weight 502.93
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.