missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ C 87
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
C 87 is a TNF-α inhibitor (Kd = 110 nM for hTNF-α). Inhibits TNF-α-induced cytotoxicity in L929 fibroblast cells. Attenuates inflammation and increases survival in a mouse hepatitis model.
Specifications
Specifications
| Chemical Name or Material | C 87 |
| CAS | 332420-90-3 |
| Synonym | 3-Phenyl-1-(4-phenyl-2-thiazolyl)-1H-pyrazole-4,5-dione 4-[2-(4-chloro-3-nitrophenyl)hydrazone] |
| Quantity | 10 mg |
| Solubility | Soluble to 20mM in DMSO |
| Molecular Formula | C24H15ClN6O3S |
| Formula Weight | 502.93 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction