missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DSR 6434
GREENER_CHOICE

Product Code. p-7082169
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18731548

Brand: Tocris Bioscience™ 4809/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent TLR7 agonist

DSR 6434 is a potent TLR7 agonist (EC50 = 7.2 nM). Activates several immune effector cells in vitro. Suppresses metastasis in vivo and enhances efficacy of radiation therapy in a model of colorectal carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DSR 6434
CAS 1059070-10-8
Synonym 6-Amino-2-(butylamino)-9-[[6-[2-(dimethylamino)ethoxy]-3-pyridinyl]methyl]-7,9-dihydro-8H-purin-8-one
Quantity 10 mg
Solubility Soluble to 50mM in 1eq. HCl and to 100mM in DMSO
Molecular Formula C19H28N8O2
Formula Weight 400.48
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.