missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GlyH 101

Description
GlyH 101 is a reversible, voltage-dependent CFTR chloride channel blocker (Ki = 4.3 μM). Inhibits forskolin -induced hyperpolarization in nasal potential differences and inhibits cholera toxin-induced intestinal fluid secretion in mice.
Specifications
Specifications
| Chemical Name or Material | GlyH 101 |
| CAS | 328541-79-3 |
| Synonym | N-2-Naphthalenyl-2-[(3,5-dibromo-2,4-dihydroxyphenyl)methylene]glycine hydrazide |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO |
| Molecular Formula | C19H15Br2N3O3 |
| Formula Weight | 493.15 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?