missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GSK LSD 1 Dihydrochloride

Description
GSK LSD 1 dihydrochloride is a potent and selective lysine specific demethylase 1 (LSD1) inhibitor (IC50 = 16 nM). Exhibits >1000-fold selectivity for LSD1 over closely related enzymes including LSD2, MAO-1 and MAO-B. Inhibits cancer cell line growth in vitro.
Specifications
Specifications
| Chemical Name or Material | GSK LSD 1 dihydrochloride |
| CAS | 2102933-95-7 |
| Synonym | rel-N-[(1S,2R)-2-Phenylcyclopropyl]-4-piperidinamine dihydrochloride |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in water and to 100mM in DMSO |
| Molecular Formula | C14H20N2·2HCl |
| Formula Weight | 289.24 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?