missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human 5-HT2B (AAH63123) VersaClone cDNA

Product Code. 18472620
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18472620 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18472620 Supplier R&D Systems™ Supplier No. RDC0289

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-Hydroxytryptamine Receptor 2A

The Human 5-HT2B (AAH63123) VersaClone cDNA clone contains the complete ORF for 5-HT2B, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The 5-HT2B insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Información de órdenes

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Especificaciones

Gene h5-HT2B
Accession Number AAH63123
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 1459bp
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg

For Reasearch Use Only; Not for Use in Humans

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.