Learn More
R&D Systems™ Human Fibronectin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 1918-FN-02M
Encompass
Product Code. 13410234
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The Human Fibronectin Protein is derived from Human Plasma. The Human Fibronectin Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Supplied as a 0.2μm filtered solution in Tris-HCl, NaCl and Urea.
Fibronectin
Human plasma-derived Fibronectin protein The human plasma used for the isolation of this product was certified by the supplier to be HIV-1 and HBsAg negative at the time of shipment. Human blood products should always be treated in accordance with universal handling precautions.
Unconjugated
2335
2mg
6 months from date of receipt, 2 to 8° C as supplied. 3 months, 2 to 8° C under sterile conditions after opening. Avoid vortexing and excessive agitation.