Learn More
R&D Systems™ Human IL-17E/IL-25 Recombinant Protein, (Human Cell-expressed)
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 8134-IL-025
Description
The Recombinant Human IL-17E (Human Cell-expressed) Protein is derived from HEK293. The Recombinant Human IL-17E (Human Cell-expressed) Protein has been validated for the following applications: Bioactivity.Specifications
| Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. | |
| 17 kDa (monomer) | |
| 25μg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
| Unconjugated |
| 64806 | |
| IL-17E/IL-25 | |
| Human embryonic kidney cell,HEK293-derived human IL-17E/IL-25 protein Tyr33-Gly177 | |
| Disulfide-linked homodimer | |
| Yes |