missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human/Mouse/Rat Activin A Protein

Product Code. 18779716
Change view
Click to view available options
Quantity:
1 mg
10 μg
50 μg
Formulation:
Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein.
Unit Size:
10µg
1mg
50µg
Product Code. Quantity Formulation unitSize
18779716 1 mg Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein. 1mg
18177420 10 μg Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein. 10µg
18757186 1 mg Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA. 1mg
18375577 50 μg Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA. 50µg
18126750 50 μg Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein. 50µg
18117780 10 μg Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA. 10µg
6 options
This item is not returnable. View return policy

Product Code. 18779716

Brand: R&D Systems™ 338AC01M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CHO derived

Measured by its ability to induce hemoglobin expression in K562 human chronic myelogenous leukemia cells. Schwall, R.H. et al. (1991) Method Enzymol. 198:340. The ED50 for this effect is 0.2-1.2 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08476
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein.
Gene ID (Entrez) 3624
Molecular Weight (g/mol) M.W. (Observed): 14 kDa, reducing conditions 24 kDa, non-reducing conditions; M.W. (theoretical): 13 kDa (monomer)
Name Activin A
Quantity 1 mg
Source Chinese Hamster Ovary cell line, CHO-derived Activin A protein Gly311-Ser426
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol INHBA
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100-500 μg/mL in sterile 4 mM HCl.
Purity or Quality Grade >95%, by SDS-PAGE with silver staining.
Protein Activin A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.