Learn More
R&D Systems™ Human TGF-beta 1 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 240-B-001MG/CF
Description
Measured by its ability to inhibit the IL-4-dependent proliferation of HT-2 mouse T cells. Tsang, M. et al. (1995) Cytokine 7:389. The ED50 for this effect is 0.04-0.2 ng/mL. The specific activity of Recombinant Human TGF-beta 1 is approximately 2.5 x 104 U/μg, which is calibrated against human TGF-beta 1 Standard (NIBSC code: 89/514). Specific activity is for reference purposes only and is not routinely tested.Specifications
| Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. | |
| 12.8 kDa (monomer) | |
| 1mg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
| CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 | |
| Yes |
| 7040 | |
| TGF-beta 1 | |
| Chinese Hamster Ovary cell line,CHO-derived human TGF-beta 1 protein Ala279-Ser390 | |
| Disulfide-linked homodimer | |
| Unconjugated |