missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human VEGFR3/Flt-4 (NP_891555) VersaClone cDNA

Product Code. 18768548
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18768548 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18768548 Supplier R&D Systems™ Supplier No. RDC1286
  • ONLINE EXCLUSIVE PRICE
    1240.00 €
    1170.75 € / 10µg
    Save 69.25 €
    5% Off
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Vascular Endothelial Growth Factor Receptor 3

The Human VEGFR3/Flt-4 (NP_891555) VersaClone cDNA clone contains the complete ORF for VEGFR3/Flt-4, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The VEGFR3/Flt-4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Product Type cDNA
Description Gene ID (Entrez): 2324, 14257
Gene hFLT4
Accession Number NP_891555
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 4105bp
Gene Alias EC 2.7.10, EC 2.7.10.1, FLT4, FLT-4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGF R3, VEGFR-3, VEGFR3Fms-like tyrosine kinase 4
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human, Mouse
Quantity 10 μg
Show More Show Less

For Research Use Only; Not for Use in Humans

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.