missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ JTE 607 Dihydrochloride
GREENER_CHOICE

Product Code. p-7082174
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18766658

Brand: Tocris Bioscience™ 5185/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cytokine release inhibitor; anti-inflammatory

JTE 607 dihydrochloride is a pro-drug that is cleaved by carboxylesterase 1 (CES1) to its active metabolite, which then binds to cleavage and polyadenylation specificity factor 3 (CPSF3; Kd = 370 nM at human CPSF3). Treatment of A-673 cells with JTE 607 results in inhibition of mRNA cleavage and accumulation of nuclear R-loops. JTE 607 induces apoptosis in leukemia cells in vitro and prolongs survival in a mouse leukemia model. JTE 607 inhibits cytokine release and inhibits production of IL-1β, IL-8, IL-6, IL-10 and TNFα (IC50 values are 5.9, 7.3, 8.8, 9.1 and 11.0 nM, respectively) from LPS-stimulated PBMCs. In a rat model of lung injury, JTE 607 reduces proinflammatory cytokine-release and attenuates lung permeability.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material JTE 607 dihydrochloride
CAS 188791-09-5
Synonym N-[3,5-Dichloro-2-hydroxy-4-[2-(4-methyl-1-piperazinyl)ethoxy]benzoyl]-L-phenylalanine ethyl ester dihydrochloride
Quantity 50 mg
Solubility Soluble to 100mM in water and to 100mM in DMSO
Molecular Formula C25H31Cl2N3O5·2HCl
Formula Weight 597.36
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.