Learn More
R&D Systems™ MagCellect Human CD4+ CD25+ Regulatory T Cell Isolation Kit
Product Code. 18451980
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. 18451980
Brand: R&D Systems™ MAGH104
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed to isolate human CD4+CD25+ regulatory T cells using a two-step procedure that combines both negative and positive selection techniques
The resulting cell preparation is highly enriched for CD4+CD25+ T cells with a purity of recovered cells ranging between 85-95%
Two-step procedure
Includes Streptavidin Ferrofluid for both the negative selection step to isolate CD4+ T cells, and in the positive selection step to isolate CD4+CD25+ regulatory T cells
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit