Learn More
Tocris Bioscience™ ML 193
Potent, selective GPR55 antagonist
Brand: Tocris Bioscience™ 4860/10
Description
ML 193 is a selective GPR55 antagonist (IC50 = 221 nM). Exhibits >27-, >145- and >145-fold selectivity for GPR55 over CB1, GPR35 and CB2, respectively. Inhibits ERK signaling in vitro.Specifications
| ML 193 | |
| N-[4-[[(3,4-Dimethyl-5-isoxazolyl)amino]sulfonyl]phenyl]-6,8-dimethyl-2-(2-pyridinyl)-4-quinolinecarboxamide | |
| Soluble to 100mM in DMSO | |
| Bioactive Small Molecule |
| 713121-80-3 | |
| 10 mg | |
| Solid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research Use Only. Not For Human or Veterinary Use