missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 193
GREENER_CHOICE

Product Code. 18784059
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18784059

Brand: Tocris Bioscience™ 4860/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, selective GPR55 antagonist

ML 193 is a selective GPR55 antagonist (IC50 = 221 nM). Exhibits >27-, >145- and >145-fold selectivity for GPR55 over CB1, GPR35 and CB2, respectively. Inhibits ERK signaling in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material ML 193
CAS 713121-80-3
Synonym N-[4-[[(3,4-Dimethyl-5-isoxazolyl)amino]sulfonyl]phenyl]-6,8-dimethyl-2-(2-pyridinyl)-4-quinolinecarboxamide
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C28H25N5O4S
Formula Weight 527.59
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.