missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse EMP2 (NP_031955) VersaClone cDNA

Produktkod. 18471330
Change view
Klicka för att se tillgängliga alternativ
Quantity:
10 μg
Förpackningsstorlek:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Förpackningsstorlek
18471330 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18471330 Leverantör R&D Systems™ Leverantörsnummer RDC0782
  • ONLINE EXCLUSIVE PRICE
    341.00 €
    322.35 € / 10µg
    Save 18.65 €
    5% Off
 Restricted Item
Estimated Shipment: 14-10-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Epithelial Membrane Protein 2

The Mouse EMP2 (NP_031955) VersaClone cDNA clone contains the complete ORF for EMP2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The EMP2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Beställningsinfo

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifikationer

Gene mEMP2
Accession Number NP_031955
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 532bp
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg

For Reasearch Use Only; Not for Use in Humans

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.