missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-4 ELISpot Kit

Product Code. p-4765397
Change view
Click to view available options
Quantity:
1 x 96-well Plate
6 x 96-well Plates
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18792164 1 x 96-well Plate 1 set
18792024 6 x 96-well Plates 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18792164 Supplier R&D Systems™ Supplier No. EL404

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

  • Microplate
  • Biotinylated Detection Antibody
  • Streptavidin conjugated to Alkaline Phosphatase
  • Dilution Buffers
  • Wash Buffer Concentrate
  • BCIP/NBT Chromogen
  • Positive Control

highly sensitive, microplate-based assays for the detection of cytokine secreting cells

Product Features:
  • Detect and quantitate individual cells secreting
  • High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
  • No in vitro expansion of cells required
  • High-throughput - ELISpot assays use only a small number of primary cells
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-4
Cross Reactivity Mouse
Includes Mouse IL-4 Microplate, Biotinylated Detection Antibody, Streptavidin conjugated to Alkaline Phosphatase, Dilution Buffers, Wash Buffer Concentrate, BCIP/NBT Chromogen, Mouse IL-4 Positive Control
Quantity 1 x 96-well Plate
Sample Type Whole cells
Synonym B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
For Use With (Application) Detection of cytokine secreting cells, quantitative sandwich ELISA
Product Type Mouse IL-4 ELISpot Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.