missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Inhibin A Protein

Product Code. 18757626
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in HCl with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in HCl.
Unit Size:
10µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation Unit Size
18757626 Lyophilized from a 0.2 μm filtered solution in HCl. 10µg
18748026 Lyophilized from a 0.2 μm filtered solution in HCl with BSA as a carrier protein. 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18757626 Supplier R&D Systems™ Supplier No. 8346IN010/CF
  • ONLINE EXCLUSIVE PRICE
    441.98 €
    393.75 € / 10µg
    Save 48.23 €
    10% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to neutralize Activin-mediated erythroid differentiation of K562 human chronic myelogenous leukemia cells. The ED50 for this effect is 5-30 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number CAA49324 (alpha chain) & P08476 (beta chain)
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in HCl.
Gene ID (Entrez) 3623
Molecular Weight (g/mol) M.W. (Observed): 12-14 kDa & 17-20 kDa, reducing conditions; M.W. (theoretical): 14.5 kDa (alpha chain) & 13.0 kDa (beta chain)
Name Inhibin A
Preparation Method Reconstitute at 100μg/mL in 4mM HCl.
Quality Control Testing SDS-PAGE: 12-14kDa & 17-20kDa, reducing conditions
Quantity 10 μg
Source Chinese Hamster Ovary cell line, CHO-derived mouse Inhibin A protein Ser234-Ile366 (alpha chain) & Gly311-Ser426 (beta chain)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Ser234 (α chain) & Gly311 (β chain)
Without Additives Carrier Protein
Structural Form Disulfide-linked heterodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Inha
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in 4 mM HCl.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Inhibin alpha
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.