Learn More
R&D Systems™ Recombinant Mouse Inhibin A Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 8346-IN-010
Description
Measured by its ability to neutralize Activin-mediated erythroid differentiation of K562 human chronic myelogenous leukemia cells. The ED50 for this effect is 5-30 ng/mL.Specifications
| CAA49324 (alpha chain) & P08476 (beta chain) | |
| 3623 | |
| Inhibin A | |
| SDS-PAGE: 12-14kDa & 17-20kDa, reducing conditions | |
| Chinese Hamster Ovary cell line, CHO-derived mouse Inhibin A protein Ser234-Ile366 (alpha chain) & Gly311-Ser426 (beta chain) | |
| RUO | |
| Disulfide-linked heterodimer | |
| Inha | |
| Yes |
| Lyophilized from a 0.2 µm filtered solution in HCl with BSA as a carrier protein. | |
| M.W. (Observed): 12-14 kDa & 17-20 kDa, reducing conditions; M.W. (theoretical): 14.5 kDa (alpha chain) & 13.0 kDa (beta chain) | |
| Reconstitute at 100μg/mL in 4mM HCl. | |
| 10 µg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| Ser234 (α chain) & Gly311 (β chain) | |
| <0.10 EU / 1 µg of the protein by the LAL method. | |
| Unconjugated | |
| Reconstitute at 100 µg/mL in 4 mM HCl. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.