missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NK 252

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
NK 252 is an Nrf2 activator; interacts with the Nrf2-binding site of Keap1. Downregulates the expression of fibrogenic genes and increases antioxidant effects in NASH rat models of liver disease.
Specifications
Specifications
| Chemical Name or Material | NK 252 |
| CAS | 1414963-82-8 |
| Synonym | N-[5-(2-Furanyl)-1,3,4-oxadiazol-2-yl]-N'-(2-pyridinylmethyl)urea |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in 1eq. HCl and to 100mM in DMSO |
| Molecular Formula | C13H11N5O3 |
| Formula Weight | 285.26 |
| Physical Form | Powder |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion