missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Vancomycin HCl

Product Code. 18791399
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18791399 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18791399 Supplier R&D Systems™ Supplier No. 5506/1G
  • ONLINE EXCLUSIVE PRICE
    281.35 €
    253.05 € / 1g
    Save 28.30 €
    10% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Antibiotic

Vancomycin HCl is an antibiotic with activity against Gram-positive bacteria.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vancomycin HCl
CAS 1404-93-9
Quantity 1 g
Solubility Soluble to 25mM in water
Product Line Tocris
Molecular Formula C66H75Cl2N9O24·HCl
Formula Weight 1485.71
Physical Form Solid
Product Type Bioactive Small Molecule

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.