missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VU 0422288

Description
VU 0422288 is a group III mGlu receptor positive allosteric modulator (EC50 values are 108, 125 and 146 nM for mGlu4, mGlu8 and mGlu7 respectively). Exhibits selectivity over a panel of 68 other GPCRs, ion channels and transporters. Potentiates mGlu7-mediated reductions in excitatory post synaptic potentials in hippocampal slices.
Specifications
Specifications
| Chemical Name or Material | VU 0422288 |
| CAS | 1630936-95-6 |
| Synonym | N-[3-Chloro-4-[(5-chloro-2-pyridinyl)oxy]phenyl]-2-pyridinecarboxamide |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO |
| Molecular Formula | C17H11Cl2N3O2 |
| Formula Weight | 360.19 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?