missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Zosuquidar Trihydrochloride
GREENER_CHOICE

Product Code. 18790278
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18790278 10 mg 10mg
18784709 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18790278 Supplier Tocris Bioscience™ Supplier No. 5456/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High affinity P-gp inhibitor

Zosuquidar trihydrochloride is a high affinity P-glycoprotein (P-gp) inhibitor (Kd = 79 nM). Restores doxorubicin (Cat. No. 2252) sensitivity in P-gp-expressing multidrug (MDR) resistant cancer cell lines. Also potentiates antitumor efficacy of taxol (Cat. No. 1097) in a MDR human non-small cell lung carcinoma xenograft mouse model.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zosuquidar trihydrochloride
CAS 167465-36-3
Synonym (αR)-4-[(1aα,6α,10bα)-1,1-Difluoro-1,1a,6,10b-tetrahydrodibenzo[a,e]cyclopropa[c]cyclohepten-6-yl]-α-[(5-quinolinyloxy)methyl]-1-piperazineethanol trihydrochloride
Quantity 10 mg
Solubility Soluble to 50mM in DMSO
Molecular Formula C32H31F2N3O2·3HCl
Formula Weight 636.99
Physical Form Solid
Product Type Bioactive Small Molecule
Purity 97%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.