missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Flurbiprofen, 97%

Product Code. 10698350 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10698350

Brand: Thermo Scientific Acros 434490010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 51543-40-9 | C15H13FO2 | 244.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 51543-40-9
CAS Min % 96.0
CAS Max % 100.0
Melting Point 106.0°C to 110.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Molecular Formula C15H13FO2
MDL Number MFCD00869714
Quantity 1 g
Merck Index 15,9196
Specific Rotation Condition − 40.00 (19.10°C c=1 in CHCl3)
Specific Rotation -40
Synonym tarenflurbil, r-flurbiprofen, flurizan, r-2-flurbiprofen, unii-501w00oowa, r-2-flubiprofen, r---flurbiprofen, --2r-2-2-fluorobiphenyl-4-yl propanoic acid, r---2-fluoro-alpha-methyl-4-biphenylacetic acid
InChI Key SYTBZMRGLBWNTM-SNVBAGLBSA-N
SMILES CC(C1=CC(=C(C=C1)C2=CC=CC=C2)F)C(=O)O
IUPAC Name (2R)-2-(3-fluoro-4-phenylphenyl)propanoic acid
Molecular Weight (g/mol) 244.27
PubChem CID 92337
ChEBI CHEBI:38666
Formula Weight 244.27
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-Flurbiprofen
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(-)-2-Fluoro-a-methyl-4-biphenylacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.