Learn More
R&D Systems™ Human GPR150 (NP_954713) VersaClone cDNA
Product Code. 18723779
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. Quantity unitSize
18723779 10 μg 10µg
1 missing translation for 'options'
Product Code. 18723779 missing translation for 'mfr' R&D Systems™ missing translation for 'supplierNo' RDC0880
G protein-coupled receptor 150
The Human GPR150 (NP_954713) VersaClone cDNA clone contains the complete ORF for GPR150, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The GPR150 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans